Learn More
Bioworld NHS lodoacetate, 500 mg
Supplier:  Bioworld 218100101
Encompass_Preferred
NHS Lodoacetate, N-Hydroxysuccinimidyl lodoacetatePrimary amine and sulfhydryl reactiveProtein-protein conjugationReferences: 1.) Rector, E.S., et al. (1978) J. Immunol. Methods. 24, 321.M.W. = 283
Catalog No. 50-199-258