Learn More
Medchemexpress LLC HY-134463 5mg Medchemexpress, NHWD-870 CAS:2115742-03-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1344635MG
Encompass_Preferred
Medchemexpress, HY-134463 5mg NHWD-870 CAS:2115742-03-3 NHWD-870 is a potent, orally active and selective BET family bromodomain inhibitor and only binds bromodomains of BRD2, BRD3, BRD4 (IC50=2.7 nM), and BRDT. NHWD-870 has potent tumor suppressive efficacies and suppresses cancer cell-macrophage interaction. NHWD-870 increases tumor apoptosis and inhibits tumor proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1366