Learn More
Cayman Chemical NNEI 2'-naphthyl isomer, Purity: ≥95%, CAS: 1338925-12-4, 10 mg
SDP
Catalog No. 502969488
Encompass_Preferred
Differs from NNEI by having the naphthalene group joined at the two position; intended for forensic and research purposes
Catalog No. 50-296-9488 Supplier Cayman Chemical Supplier No. 1473810 MG
Differs from NNEI by having the naphthalene group joined at the two position; intended for forensic and research purposes