Learn More
Medchemexpress LLC HY-19537 5mg Medchemexpress, NI-57 CAS:1883548-89-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY195375MG
Encompass_Preferred
Medchemexpress, HY-19537 5mg NI-57 CAS:1883548-89-7 NI-57 is an inhibitor of bromodomain and plant homeodomain finger-containing (BRPF) famlily of proteins, with IC50s of 3.1, 46 and 140 nM for BRPF1, BRPF2 (BRD1) and BRPF3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2140