Learn More
Gold Biotechnology Inc Nickel NTA Magnetic Agarose Beads 2 mL
SDP
Catalog No. NC2601405
Encompass
Nickel NTA Magnetic Agarose Beads are a rapid and easy small scale purification of histidine-tagged proteins. This resin consists of magnetic agarose derivatized with Nitrilotriacetic (NTA) and provides good properties working in native or denaturing conditions. Magnetic agarose beads provide a convenient and quick method for purification without the need for pipetting or centrifugation. Washing, binding and elution steps will require a magnetic device.
Catalog No. NC2601405 Supplier Gold Biotechnology Inc Supplier No. H3512
Nickel NTA Magnetic Agarose Beads are a rapid and easy small scale purification of histidine-tagged proteins. This resin consists of magnetic agarose derivatized with Nitrilotriacetic (NTA) and provides good properties working in native or denaturing conditions. Magnetic agarose beads provide a convenient and quick method for purification without the need for pipetting or centrifugation. Washing, binding and elution steps will require a magnetic device.