Learn More
Genscript Corporation Ni-charged MagBeads
Catalog No. NC1191649
Encompass
The GenScript Ni-Charged MagBeads are developed for quick and efficient small-scale purification of polyhistidine-tagged proteins under native or denaturing conditions.Perform cell lysis under native or denaturing conditions. Add the cell lysate containing your polyhistidine-tagged proteins to the Ni-Charged MagBeads and allow the proteins bind to the MagBeads. Then the isolated proteins can be eluted from the beads. Magnetic separation eliminates the need for multiple tubes, minimizes the loss of sample and removes several steps of the centrifugation process.
Catalog No. NC1191649 Supplier Genscript Corporation Supplier No. L00295
The GenScript Ni-Charged MagBeads are developed for quick and efficient small-scale purification of polyhistidine-tagged proteins under native or denaturing conditions.Perform cell lysis under native or denaturing conditions. Add the cell lysate containing your polyhistidine-tagged proteins to the Ni-Charged MagBeads and allow the proteins bind to the MagBeads. Then the isolated proteins can be eluted from the beads. Magnetic separation eliminates the need for multiple tubes, minimizes the loss of sample and removes several steps of the centrifugation process.