Learn More
Bioworld NI-CHELATED SEPAROPOREAGA
Supplier:  Bioworld 201810463
Encompass
NI-CHELATED SEPAROPORE (AGAROSE) 4B: Ni-Chelated Separopore 4B is an imminodiacetate (IDA)-immobilized metal affinity chromatography (IMAC) matrix that seperates proteins via their differences in affinity to nickel metal ions. Propertie...
Catalog No. 50-197-783