Learn More
Strem, An Ascensus Company CAS# 3264-82-2. 100G. Nickel(II) acetylacetonate, anhydrous, min. 95%. MFCD00000024
SDP
Catalog No. 501963540
Encompass
CAS# 3264-82-2. 100G. Nickel(II) acetylacetonate, anhydrous, min. 95%. MFCD00000024. Molecular Formula: Ni(CH3COCHCOCH3)2. Molecular Weight: 256.93. Color/Form: light green pwdr. Stability: hygroscopic. Strem# 28-1130. www.strem.com/catalog/v3/28-1130/
Catalog No. 50-196-3540 Supplier Strem, An Ascensus Company Supplier No. 281130100G
CAS# 3264-82-2. 100G. Nickel(II) acetylacetonate, anhydrous, min. 95%. MFCD00000024. Molecular Formula: Ni(CH3COCHCOCH3)2. Molecular Weight: 256.93. Color/Form: light green pwdr. Stability: hygroscopic. Strem# 28-1130. www.strem.com/catalog/v3/28-1130/