Learn More
Bioworld Ni-IMAC 5 x 2mL Columns, 5 Column
Catalog No. 50197626
Encompass
Ni-immobilized metal affinity chromatography (IMAC) columns consist of Ni-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Ni-IMAC Columns separate proteins via their differences in affinity to nickel metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged with Ni metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 6BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Ni2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.
Catalog No. 50-197-626 Supplier Bioworld Supplier No. 201700061
Ni-immobilized metal affinity chromatography (IMAC) columns consist of Ni-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Ni-IMAC Columns separate proteins via their differences in affinity to nickel metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged with Ni metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 6BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Ni2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.