Learn More
CUBE BIOTECH PureCube Ni-NTA Agarose; 1000 ml
Catalog No. NC2684630
Encompass
1 L Ni-NTA MagBeads for His-tag-purification, 25% slurry
Catalog No. NC2684630 Supplier CUBE BIOTECH Supplier No. 31115
Add to Cart
Edge
Add to Cart
Request a Quote
1 L Ni-NTA MagBeads for His-tag-purification, 25% slurry