Learn More
CUBE BIOTECH PureCube Ni-NTA MagBeads; 25 ml
Catalog No. NC2684632
Encompass
25 ml Ni-NTA MagBeads for His-tag-purification, 25% slurry
Catalog No. NC2684632 Supplier CUBE BIOTECH Supplier No. 31225
25 ml Ni-NTA MagBeads for His-tag-purification, 25% slurry