Learn More
Medchemexpress LLC Ni-NTA Magnetic Agarose Beads | 1 ML
SDP
Catalog No. 5000428849
Encompass_Preferred
MCE Ni-NTA Magnetic Agarose Beads are highly cross-linked 4% magnetic agarose, synthesized through the chemically directed, high-density attachment of nitrilotriacetic acid (NTA). When chelated with Ni2+, it forms an extremely stable planar tetragonal structure, providing three ionic bonding sites. It has high loading capacity, exceptional specificity, and stability. The product can be used for the detection and purification of His-tag proteins from various expression sources such as E.coli, yeast, insect cells, and mammalian cells. The specifications of the product correspond to the actual beads volume, with the beads content of 50%.
  • High loading capacity
  • Exceptional specificity
  • High stability
  • Can be used for the detection and purification of His-tag proteins from various expression sources (E.coli, yeast, insect cells, and mammalian cells)
Catalog No. 50-004-28849 Supplier Medchemexpress LLC Supplier No. HYK02411ML
MCE Ni-NTA Magnetic Agarose Beads are highly cross-linked 4% magnetic agarose, synthesized through the chemically directed, high-density attachment of nitrilotriacetic acid (NTA). When chelated with Ni2+, it forms an extremely stable planar tetragonal structure, providing three ionic bonding sites. It has high loading capacity, exceptional specificity, and stability. The product can be used for the detection and purification of His-tag proteins from various expression sources such as E.coli, yeast, insect cells, and mammalian cells. The specifications of the product correspond to the actual beads volume, with the beads content of 50%.
  • High loading capacity
  • Exceptional specificity
  • High stability
  • Can be used for the detection and purification of His-tag proteins from various expression sources (E.coli, yeast, insect cells, and mammalian cells)