Learn More
New England Biolabs, Inc. NEBExpress Ni-NTA Magnetic Beads – 1 ml
SDP
Supplier:  New England Biolabs, Inc. S1423S
Encompass
An affinity matrix for the small-scale isolation and purification of polyhistidine-tagged (His-tagged) fusion proteins in manual or automated formats. Immobilized Metal Affinity Chromatography (IMAC) purifications employing Ni-NTA (nickel-nitrilotriacetic acid) magnetic beads can be performed under native or denaturing conditions which permit efficient binding and purification of insoluble proteins, proteins that aggregate in inclusion bodies, or proteins that possess a tertiary structure that sequester the polyhistidine affinity tag.
Catalog No. 50-195-9413