Learn More
Invitrogen™ Ni-NTA Purification System
Catalog No. k95001
Encompass_Preferred
Click to view available options
Quantity:
6 purifications
Includes
6 x 2mL resin columns and buffers for native and denaturing purification; 10mL Ni-NTA Agarose.
Designed for the purification of recombinant proteins that contain a polyhistidine (6xHis) sequence
  • Utilizes Ni-NTA nickel-chelating resin and is supplied with native and denaturing buffers for efficient purification of recombinant proteins under different conditions
His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Format Suspension
Product Type Ni-NTA Purification System
Content And Storage Six 2-ml resin columns and buffers for native and denaturing purification. Ten milliliters of Ni-NTA Agarose. Store at +4°C. All reagents are guaranteed stable for 6 months when properly stored.
Protein Tag His Tag
Quantity 6 purifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.