Learn More
MARVELGENT BIOSCIENCES INC Ni-Penta Agarose, 100 mL
SDP
Catalog No. NC1210120
Encompass_Preferred
Ni-Penta Agarose is a new immobilized metal ion affinity chromatography (IMAC) medium precharged with nickel ions, designed mainly for purification of low abundant proteins or challenging proteins in conditions where EDTA, DTT, beta-ME, and other stabilizing agents are necessary for maintaining protein stability and integrity.
Catalog No. NC1210120 Supplier MARVELGENT BIOSCIENCES INC Supplier No. 110227100
Ni-Penta Agarose is a new immobilized metal ion affinity chromatography (IMAC) medium precharged with nickel ions, designed mainly for purification of low abundant proteins or challenging proteins in conditions where EDTA, DTT, beta-ME, and other stabilizing agents are necessary for maintaining protein stability and integrity.