Learn More
MARVELGENT BIOSCIENCES INC PROTEINDEX™ Ni-Penta™ Agarose 6 Fast Flow, Chemical Stable, Prepacked Cartridge 5x5mL
SDP
Catalog No. NC1376357
Encompass_Preferred
PROTEINDEX™ Chemical-Tolerant Ni-Penta™ Agarose 6FF is a new immobilized metal ion affinity chromatography (IMAC) medium precharged with nickel ions, designed mainly for purification of low abundant proteins or challenging proteins in conditions where EDTA, DTT, β-ME, and other stabilizing agents are necessary for maintaining protein stability and integrity. The nickel ion binds very strongly to the matrix support in complex with a proprietary pentadentate chelating ligand, which immobilizes the nickel ion through 5 sites to form a homogeneous, chemical-stable structure. This characteristic nickel ion bonding on the Ni-Penta™ resin provides excellent resistance to EDTA, NaOH, β-ME, and other reagents that are restricted in the purification protocol using conventional NTA- and IDA- functionalized resins.
Catalog No. NC1376357 Supplier MARVELGENT BIOSCIENCES INC Supplier No. 1102295X5ML
PROTEINDEX™ Chemical-Tolerant Ni-Penta™ Agarose 6FF is a new immobilized metal ion affinity chromatography (IMAC) medium precharged with nickel ions, designed mainly for purification of low abundant proteins or challenging proteins in conditions where EDTA, DTT, β-ME, and other stabilizing agents are necessary for maintaining protein stability and integrity. The nickel ion binds very strongly to the matrix support in complex with a proprietary pentadentate chelating ligand, which immobilizes the nickel ion through 5 sites to form a homogeneous, chemical-stable structure. This characteristic nickel ion bonding on the Ni-Penta™ resin provides excellent resistance to EDTA, NaOH, β-ME, and other reagents that are restricted in the purification protocol using conventional NTA- and IDA- functionalized resins.