Learn More
Genscript Corporation Ni Resin FF
Catalog No. 5000058949
Encompass
GenScript Ni Resin FF is a 6% highly cross-linked agarose medium covalently coupled to a chelating agent that binds Ni sup 2 /sup by three coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins
Catalog No. 50-000-58949 Supplier Genscript Corporation Supplier No. L0046525
GenScript Ni Resin FF is a 6% highly cross-linked agarose medium covalently coupled to a chelating agent that binds Ni sup 2 /sup by three coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins