Learn More
Genscript Corporation Ni Resin FF
Supplier:  Genscript Corporation L004655
Encompass
GenScript Ni Resin FF is a 6% highly cross-linked agarose medium covalently coupled to a chelating agent that binds Ni sup 2 /sup by three coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins
Catalog No. 50-000-58950