Learn More
Glass Expansion Nickel Sampler Cone with Copper Core for Thermo Scientific X/Q/RQ/TQ/MX Series ICP-MS. Includes graphite gasket. (3600812)
SDP
Catalog No. 501694614
Encompass_Preferred
Nickel Sampler Cone with Copper Core (3600812), standard sampler cone for Thermo Q/RQ/TQ ICP-MS. Optional sampler cone with more efficient heat transfer for Thermo X Series ICP-MS. Suitable for most common applications (e.g., routine aqueous environmental samples, <5% acid matrix, non-HF, non-organic). Use a new Graphite Sampler Cone Gasket (TG5001) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.As acid concentrations begin to exceed 5%, and for samples containing hydrofluoric acid (HF), we recommend platinum cones. For samples containing HF and other highly corrosive samples, see catalog number TG1026A-Pt/Cu. For lowest Ni background and laser ablation applications see catalog number TG1021-Al.
Catalog No. 50-169-4614 Supplier Glass Expansion Supplier No. TG1021NI/CU
Nickel Sampler Cone with Copper Core (3600812), standard sampler cone for Thermo Q/RQ/TQ ICP-MS. Optional sampler cone with more efficient heat transfer for Thermo X Series ICP-MS. Suitable for most common applications (e.g., routine aqueous environmental samples, <5% acid matrix, non-HF, non-organic). Use a new Graphite Sampler Cone Gasket (TG5001) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.As acid concentrations begin to exceed 5%, and for samples containing hydrofluoric acid (HF), we recommend platinum cones. For samples containing HF and other highly corrosive samples, see catalog number TG1026A-Pt/Cu. For lowest Ni background and laser ablation applications see catalog number TG1021-Al.