Learn More
Strem, An Ascensus Company CAS #2468315-70-8. 1g. Tris(trans-1,2-bis(4-tert-butylphenyl)ethene)nickel(0), min. 97%
SDP
Supplier:  Strem, An Ascensus Company 2800701G
Encompass
CAS #2468315-70-8. 1g. Tris(trans-1,2-bis(4-tert-butylphenyl)ethene)nickel(0), min. 97%. Molecular Weight: 936.07. Molecular Formula: C66H84Ni. Color/form: red-orange pwdr. Strem# 28-0070. http://www.strem.com/catalog/v/28-0070/
Catalog No. 50-204-5286