Learn More
WELCH MATERIALS INC Ni TanroseTM 6FF(NTA), 100mL/bottle
Catalog No. 502093191
Encompass_Preferred
Name: Ni Tanrose 6FF(NTA), specification: 100mL, bead structure: 6% highly cross-linked agarose, mean particle size and range: 90μm, 45-165 μm, binding capacity: 40mg(his-tagged protein)/mL(medium), pH stability: 3-12(long term), 2-14(short term), flow rate: 600 cm/h, storage condition: 20% ethanol, 4-30°C. Similar Products: Ni Sepharose 6 FF
Catalog No. 50-209-3191 Supplier WELCH MATERIALS INC Supplier No. W5016
Name: Ni Tanrose 6FF(NTA), specification: 100mL, bead structure: 6% highly cross-linked agarose, mean particle size and range: 90μm, 45-165 μm, binding capacity: 40mg(his-tagged protein)/mL(medium), pH stability: 3-12(long term), 2-14(short term), flow rate: 600 cm/h, storage condition: 20% ethanol, 4-30°C. Similar Products: Ni Sepharose 6 FF