Learn More
Medchemexpress LLC Niacin-13C6 | 1189954-79-7 | 99.9% | 1 MG
SDP
Catalog No. 5000450473
Encompass_Preferred
Niacin-13C6 is the 13C-labeled Niacin. Niacin (Nicotinic acid) is a vitamin and is part of the vitamin B group.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-50473 Supplier Medchemexpress LLC Supplier No. HYB0143S31MG
Niacin-13C6 is the 13C-labeled Niacin. Niacin (Nicotinic acid) is a vitamin and is part of the vitamin B group.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.