Learn More
Medchemexpress LLC Niacin-d4 | 66148-15-0 | 99.7% | 5 MG
SDP
Catalog No. 5000450919
Encompass_Preferred
Niacin-d4 is the deuterium labeled Niacin, a vitamin and part of the vitamin B group. It is intended for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50919 Supplier Medchemexpress LLC Supplier No. HYB0143S25MG
Niacin-d4 is the deuterium labeled Niacin, a vitamin and part of the vitamin B group. It is intended for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS