Learn More
Medchemexpress LLC Niacin-d4 | 66148-15-0 | MFCD00190491 | 99.6% | 1 MG
SDP
Catalog No. 5000450918
Encompass_Preferred
Niacin-d4 is the deuterium labeled Niacin. Niacin (Nicotinic acid) is a vitamin and is part of the vitamin B group. It is intended for research use only. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Niacin.
  • Vitamin B group member.
  • Suitable for research use.
  • Functions as a tracer or internal standard.
  • Can be used in quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence pharmacokinetic and metabolic profiles.
Catalog No. 50-004-50918 Supplier Medchemexpress LLC Supplier No. HYB0143S21MG
Niacin-d4 is the deuterium labeled Niacin. Niacin (Nicotinic acid) is a vitamin and is part of the vitamin B group. It is intended for research use only. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Niacin.
  • Vitamin B group member.
  • Suitable for research use.
  • Functions as a tracer or internal standard.
  • Can be used in quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence pharmacokinetic and metabolic profiles.