Learn More
Medchemexpress LLC Niacin (standard) | 59-67-6 | 100.0% | 250 MG
SDP
Catalog No. 5000407046
Encompass_Preferred
Niacin (Standard) serves as the analytical standard for Niacin and is intended for research and analytical applications. It is an orally active, water-soluble B3 vitamin that is essential for human health. Niacin (Vitamin B3) is crucial for energy metabolism, cell signaling pathways that regulate gene expression and apoptosis, and is also utilized in cardiovascular disease research.
  • Grade: Analytical standard, supplied assay reference standard.
  • Commonly used in: Qualitative, quantitative, and methodological research experiments.
  • Analytical techniques: Suitable for HPLC, GC, and MS.
Catalog No. 50-004-07046 Supplier Medchemexpress LLC Supplier No. 250MGHYB0143R
Add to Cart
Add to Cart
Niacin (Standard) serves as the analytical standard for Niacin and is intended for research and analytical applications. It is an orally active, water-soluble B3 vitamin that is essential for human health. Niacin (Vitamin B3) is crucial for energy metabolism, cell signaling pathways that regulate gene expression and apoptosis, and is also utilized in cardiovascular disease research.
  • Grade: Analytical standard, supplied assay reference standard.
  • Commonly used in: Qualitative, quantitative, and methodological research experiments.
  • Analytical techniques: Suitable for HPLC, GC, and MS.