Learn More
U.S. Pharmacopeia Niacinamide, 98-92-0, MFCD00006395, 500mg
Catalog No. NC9598089
Encompass_Preferred
Molecular formula C6-H6-N2-O, Molecular weight 122.13 g/mol, Melting Point 262.4 - 267.8 °F (128 - 131 °C), Boiling Point 314.6 °F (157 °C), Synonyms: Nicotinamide, Vitamin B3
Catalog No. NC9598089 Supplier U.S. Pharmacopeia Supplier No. 1462006
Molecular formula C6-H6-N2-O, Molecular weight 122.13 g/mol, Melting Point 262.4 - 267.8 °F (128 - 131 °C), Boiling Point 314.6 °F (157 °C), Synonyms: Nicotinamide, Vitamin B3