Learn More
Apexbio Technology LLC Nicotinamide Riboside Chloride (NIAGEN)(Synonyms: NR Chloride, Niagen, Nicotinamide Riboside Chloride, NR-Cl, Nicotinamide Riboside Cl), 200mg, CAS: 23111-00-4.
SDP
Catalog No. 5000061250
Encompass_Preferred
Nicotinamide Riboside Chloride (NIAGEN CAS 23111-00-4) is a small molecule precursor of nicotinamide adenine dinucleotide (NAD ) an essential cofactor implicated in energy metabolism and cellular homeostasis Upon oral administration it effectively elevates intracellular NAD levels subsequently modulating the activity of NAD -dependent sirtuin enzymes particularly SIRT1 and SIRT3 Experimental evidence indicates that Nicotinamide Riboside Chloride enhances oxidative metabolism and mitigates metabolic dysfunction induced by high-fat dietary conditions Additionally studies employing Alzheimer s disease transgenic mouse models suggest that this compound can reduce cognitive decline supporting its use in biomedical research targeting metabolic and neurodegenerative conditions
Catalog No. 50-000-61250 Supplier Apexbio Technology LLC Supplier No. C7038200
Nicotinamide Riboside Chloride (NIAGEN CAS 23111-00-4) is a small molecule precursor of nicotinamide adenine dinucleotide (NAD ) an essential cofactor implicated in energy metabolism and cellular homeostasis Upon oral administration it effectively elevates intracellular NAD levels subsequently modulating the activity of NAD -dependent sirtuin enzymes particularly SIRT1 and SIRT3 Experimental evidence indicates that Nicotinamide Riboside Chloride enhances oxidative metabolism and mitigates metabolic dysfunction induced by high-fat dietary conditions Additionally studies employing Alzheimer s disease transgenic mouse models suggest that this compound can reduce cognitive decline supporting its use in biomedical research targeting metabolic and neurodegenerative conditions