Learn More
ATCC Niallia circulans
Catalog No. 502383705
Encompass
Niallia circulans strain AMC 732 is a quality control strain that is included in the bioMerieux VITEK 2 - Bacillus (BCL) Card Quality Control Organism Set.
Catalog No. 50-238-3705 Supplier ATCC Supplier No. 61
Niallia circulans strain AMC 732 is a quality control strain that is included in the bioMerieux VITEK 2 - Bacillus (BCL) Card Quality Control Organism Set.