Learn More
Medchemexpress LLC Nibr-0213 | 1233332-14-3 | MFCD25977008 | ≥99.0% | 464.98 | C27H29ClN2O3 | 10 MG
SDP
Catalog No. 5000298581
Encompass_Preferred
NIBR-0213 is a potent, orally active, and selective sphingosine-1-phosphate receptor 1 (S1P1) antagonist used as a research compound in immunology and pharmacology studies. It exhibits low-nanomolar potency on human and rat S1P1 and has shown efficacy in preclinical models of autoimmune encephalomyelitis. Supplied as a solid with documented analytical purity suitable for biochemical and pharmacological applications.
  • Selective S1P1 antagonist for research use.
  • Demonstrated efficacy in autoimmune encephalomyelitis models.
  • Low-nanomolar potency on human and rat S1P1.
  • High documented purity for analytical assays.
  • Solid form; store sealed away from moisture.
Catalog No. 50-002-98581 Supplier Medchemexpress LLC Supplier No. HY1816610MG
NIBR-0213 is a potent, orally active, and selective sphingosine-1-phosphate receptor 1 (S1P1) antagonist used as a research compound in immunology and pharmacology studies. It exhibits low-nanomolar potency on human and rat S1P1 and has shown efficacy in preclinical models of autoimmune encephalomyelitis. Supplied as a solid with documented analytical purity suitable for biochemical and pharmacological applications.
  • Selective S1P1 antagonist for research use.
  • Demonstrated efficacy in autoimmune encephalomyelitis models.
  • Low-nanomolar potency on human and rat S1P1.
  • High documented purity for analytical assays.
  • Solid form; store sealed away from moisture.