Learn More
Medchemexpress LLC NIC-0102 | 2806031-94-5 | C21H25BF2N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1512525MG
Encompass_Preferred
NIC-0102 is an orally active proteasome inhibitor (pIC50=7.55) that specifically inhibits NLRP3 inflammatory vesicle activation. It demonstrates potent anti-inflammatory effects in a model of dextran sulfate sodium (DSS)-induced ulcerative colitis, and it also inhibits the production of pro-IL-1β. This product is for research use only.
  • Specifically inhibits NLRP3 inflammatory vesicle activation
  • Shows potent anti-inflammatory effects in DSS-induced ulcerative colitis model
  • Inhibits production of pro-IL-1β
Catalog No. 50-003-76943