Learn More
Medchemexpress LLC HY-100592 10mg Medchemexpress, Nicaraven CAS:79455-30-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10059210MG
Encompass_Preferred
Medchemexpress, HY-100592 10mg Nicaraven CAS:79455-30-4 Nicaraven is a novel chemically synthesized hydroxyl radical-specific scavenger. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1896