Learn More
Medchemexpress LLC Nicarbazin-d8 | 00-00-0 | MFCD00867204 | 100.0% | 434.43 | C19H10D8N6O6 | 1 MG
SDP
Catalog No. 5000242764
Encompass_Preferred
Nicarbazin-d8 is a deuterium-labeled analogue of nicarbazin intended for use as an analytical internal standard and tracer in drug metabolism, pharmacokinetic, and quantitative bioanalytical studies. Supplied as a white to off-white powder for research use only; not for human or veterinary use.
  • Deuterium-labeled (d8) isotopologue for clear mass shifts in mass spectrometry analyses.
  • High purity suitable for use as an internal standard in LC-MS, GC-MS, and NMR.
  • Solid powder form with defined storage conditions to preserve stability.
  • Appropriate for pharmacokinetic, metabolism, and quantitative analytical studies.
  • White to off-white color enables easy visual inspection.
  • Available in small research pack sizes for analytical workflows.
Catalog No. 50-002-42764 Supplier Medchemexpress LLC Supplier No. HY107814S1MG
Nicarbazin-d8 is a deuterium-labeled analogue of nicarbazin intended for use as an analytical internal standard and tracer in drug metabolism, pharmacokinetic, and quantitative bioanalytical studies. Supplied as a white to off-white powder for research use only; not for human or veterinary use.
  • Deuterium-labeled (d8) isotopologue for clear mass shifts in mass spectrometry analyses.
  • High purity suitable for use as an internal standard in LC-MS, GC-MS, and NMR.
  • Solid powder form with defined storage conditions to preserve stability.
  • Appropriate for pharmacokinetic, metabolism, and quantitative analytical studies.
  • White to off-white color enables easy visual inspection.
  • Available in small research pack sizes for analytical workflows.