Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicardipine-d3 hydrochloride | 1432061-50-1 | MFCD30747912 | 99.7% | 519.00 | C26H27D3ClN3O6 | 10MG
SDP

Catalog No. 5000238121
Encompass_Preferred

Nicardipine-d3 hydrochloride is a deuterium-labeled hydrochloride salt of nicardipine supplied as a high-purity solid analytical standard for research use. It is intended as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses in analytical and pharmacokinetic workflows.

  • Deuterated tracer for quantitative analysis by NMR, GC-MS, and LC-MS.
  • High purity (99.72%) suitable for analytical standard use.
  • Solid hydrochloride form for stable handling and storage.
  • Available in small packaging sizes tailored for analytical workflows.
  • Molecular weight 519.00 and chemical formula C26H27D3ClN3O6.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-38121 Supplier Medchemexpress LLC Supplier No. HY12515AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Nicardipine-d3 hydrochloride is a deuterium-labeled hydrochloride salt of nicardipine supplied as a high-purity solid analytical standard for research use. It is intended as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses in analytical and pharmacokinetic workflows.

  • Deuterated tracer for quantitative analysis by NMR, GC-MS, and LC-MS.
  • High purity (99.72%) suitable for analytical standard use.
  • Solid hydrochloride form for stable handling and storage.
  • Available in small packaging sizes tailored for analytical workflows.
  • Molecular weight 519.00 and chemical formula C26H27D3ClN3O6.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.