Learn More
Medchemexpress LLC Nicardipine-d3 hydrochloride | 1432061-50-1 | MFCD30747912 | 99.7% | 519.00 | C26H27D3ClN3O6 | 1MG
SDP
Catalog No. 5000235554
Encompass_Preferred
Nicardipine-d3 hydrochloride is the trideuterated hydrochloride salt of nicardipine provided as a research-grade isotopically labeled reference standard for analytical chemistry and mass spectrometry applications. It is intended for use as an internal standard in LC-MS method development, quantitation, and related laboratory studies. For research use only.
  • Deuterium-labeled analogue suitable as an internal standard.
  • High isotopic enrichment to improve mass-spectrometry accuracy.
  • High chemical purity appropriate for analytical workflows.
  • Hydrochloride salt form for improved stability and handling.
  • Available in milligram quantities for trace-level standard preparation.
Catalog No. 50-002-35554 Supplier Medchemexpress LLC Supplier No. HY12515AS1MG
Nicardipine-d3 hydrochloride is the trideuterated hydrochloride salt of nicardipine provided as a research-grade isotopically labeled reference standard for analytical chemistry and mass spectrometry applications. It is intended for use as an internal standard in LC-MS method development, quantitation, and related laboratory studies. For research use only.
  • Deuterium-labeled analogue suitable as an internal standard.
  • High isotopic enrichment to improve mass-spectrometry accuracy.
  • High chemical purity appropriate for analytical workflows.
  • Hydrochloride salt form for improved stability and handling.
  • Available in milligram quantities for trace-level standard preparation.