Learn More
Medchemexpress LLC HY-12515A 500mg , Nicardipine (hydrochloride) CAS:54527-84-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12515A500MG
Encompass_Preferred
HY-12515A 500mg Nicardipine (hydrochloride) CAS: 54527-84-3 Nicardipine hydrochloride (YC-93) is a calcium channel blocker with an IC50 of 1 μM for blocking cardiac calcium channels. Nicardipine hydrochloride acts as an agent for chronic stable angina and for controlling blood pressure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0005