Learn More
Apexbio Technology LLC Nicardipine HCl 54527-84-3 5g
SDP
Catalog No. 5000063762
Encompass_Preferred
Nicardipine HCl (CAS 54527-84-3) is a dihydropyridine-derived small molecule that functions primarily as a calcium-channel blocker through inhibition of voltage-dependent L-type calcium channels This mechanism prevents calcium ion influx causing vasodilation and decreased vascular resistance Widely applied in vascular biology and pharmacological research nicardipine hydrochloride serves as a useful tool to explore calcium-dependent processes cardiovascular pathophysiology and therapeutic strategies targeting calcium-mediated vascular conditions
Catalog No. 50-000-63762 Supplier Apexbio Technology LLC Supplier No. B17985000
Nicardipine HCl (CAS 54527-84-3) is a dihydropyridine-derived small molecule that functions primarily as a calcium-channel blocker through inhibition of voltage-dependent L-type calcium channels This mechanism prevents calcium ion influx causing vasodilation and decreased vascular resistance Widely applied in vascular biology and pharmacological research nicardipine hydrochloride serves as a useful tool to explore calcium-dependent processes cardiovascular pathophysiology and therapeutic strategies targeting calcium-mediated vascular conditions