Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicardipine hydrochloride | 54527-84-3 | MFCD00057327 | ≥98.0% | 515.99 g/mol | C26H30ClN3O6 | 10MM 1ML
SDP

Catalog No. 5000238755
Encompass_Preferred

Nicardipine hydrochloride is a dihydropyridine L-type calcium channel blocker supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research, including cardiac calcium channel and vascular pharmacology assays.

  • Ready-to-use 10 mM solution in DMSO for accurate dosing.
  • Supplied in a 1 mL volume suitable for small-scale assays.
  • Applicable to cardiac calcium channel and vascular smooth muscle studies.
  • High purity suitable for research (≥98% when supplied as powder).
  • Molecular formula C26H30ClN3O6; molecular weight ~516 g/mol.

Catalog No. 50-002-38755 Supplier Medchemexpress LLC Supplier No. HY12515A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicardipine hydrochloride is a dihydropyridine L-type calcium channel blocker supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research, including cardiac calcium channel and vascular pharmacology assays.

  • Ready-to-use 10 mM solution in DMSO for accurate dosing.
  • Supplied in a 1 mL volume suitable for small-scale assays.
  • Applicable to cardiac calcium channel and vascular smooth muscle studies.
  • High purity suitable for research (≥98% when supplied as powder).
  • Molecular formula C26H30ClN3O6; molecular weight ~516 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.