Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicardipine hydrochloride | 54527-84-3 | MFCD00057327 | 99.7% | 515.99 g/mol | C26H30ClN3O6 | 250MG
SDP

Catalog No. 5000238756
Encompass_Preferred

Nicardipine hydrochloride is the hydrochloride salt of nicardipine, supplied as a research-grade analytical standard for use in pharmacology, biochemical assays, and reference material applications. It functions as an L-type calcium channel blocker and is used as a calibration or reference compound in cardiovascular and ion-channel studies.

  • High purity suitable for analytical applications, 99.7%.
  • Well-characterized identity with CAS 54527-84-3 and molecular formula C26H30ClN3O6.
  • Available in multiple solid pack sizes and solution formats for assay convenience.
  • Used as a reference standard in calcium channel and pharmacology studies.
  • Stable solid form with a defined molecular weight of 515.99 g/mol.

Catalog No. 50-002-38756 Supplier Medchemexpress LLC Supplier No. HY12515AR250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicardipine hydrochloride is the hydrochloride salt of nicardipine, supplied as a research-grade analytical standard for use in pharmacology, biochemical assays, and reference material applications. It functions as an L-type calcium channel blocker and is used as a calibration or reference compound in cardiovascular and ion-channel studies.

  • High purity suitable for analytical applications, 99.7%.
  • Well-characterized identity with CAS 54527-84-3 and molecular formula C26H30ClN3O6.
  • Available in multiple solid pack sizes and solution formats for assay convenience.
  • Used as a reference standard in calcium channel and pharmacology studies.
  • Stable solid form with a defined molecular weight of 515.99 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.