Learn More
Medchemexpress LLC Nicardipine hydrochloride | 54527-84-3 | MFCD00057327 | 99.7% | 515.99 g/mol | C26H30ClN3O6 | 25MG
SDP
Catalog No. 5000238209
Encompass_Preferred
Nicardipine hydrochloride (standard) is an analytical reference standard of the calcium channel blocker nicardipine intended for research, method development, and quality control applications. The hydrochloride salt is supplied with high chromatographic purity and is packaged to preserve stability for analytical use. Store sealed, protected from light and moisture at 4°C.
Catalog No. 50-002-38209 Supplier Medchemexpress LLC Supplier No. HY12515AR25MG
Nicardipine hydrochloride (standard) is an analytical reference standard of the calcium channel blocker nicardipine intended for research, method development, and quality control applications. The hydrochloride salt is supplied with high chromatographic purity and is packaged to preserve stability for analytical use. Store sealed, protected from light and moisture at 4°C.