Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicardipine hydrochloride | 54527-84-3 | MFCD00057327 | 99.9% | 515.99 g·mol⁻¹ | C26H29N3O6 · HCl | 5G
SDP

Catalog No. 5000238490
Encompass_Preferred

Nicardipine hydrochloride is the hydrochloride salt of nicardipine, a dihydropyridine L-type (voltage-dependent) calcium channel blocker supplied as a powder for laboratory research. It is used in pharmacology and physiology studies related to calcium channels, cardiovascular function, and neurobiology, and is available in multiple pack sizes for experimental use.

  • Powder form for laboratory use.
  • High purity (≈99.85%) suitable for analytical and biological studies.
  • Molecular formula C26H29N3O6 · HCl.
  • Molecular weight 515.99 g·mol⁻¹.
  • Used as an L-type calcium channel blocker in cardiovascular and neurobiological research.
  • Available in multiple pack sizes, including 5 g.
  • Store refrigerated as reported by distributor.

Catalog No. 50-002-38490 Supplier Medchemexpress LLC Supplier No. HY12515A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicardipine hydrochloride is the hydrochloride salt of nicardipine, a dihydropyridine L-type (voltage-dependent) calcium channel blocker supplied as a powder for laboratory research. It is used in pharmacology and physiology studies related to calcium channels, cardiovascular function, and neurobiology, and is available in multiple pack sizes for experimental use.

  • Powder form for laboratory use.
  • High purity (≈99.85%) suitable for analytical and biological studies.
  • Molecular formula C26H29N3O6 · HCl.
  • Molecular weight 515.99 g·mol⁻¹.
  • Used as an L-type calcium channel blocker in cardiovascular and neurobiological research.
  • Available in multiple pack sizes, including 5 g.
  • Store refrigerated as reported by distributor.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.