Learn More
Enzo Life Sciences Nicardipine . hydrochloride (5g). CAS: 54527-84-3
SDP
Supplier:  Enzo Life Sciences ALX550273G005
Encompass_Preferred
Ca2+ channel antagonist. Antihypertensive. Alternative name: YC-93 . HCl, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-[methyl(phenylmethyl)amino]ethyl ester . HCl. Purity: ≥98% (HPLC). Appearance: Light yellow solid. Solubility: Soluble in DMSO (1mg/ml), ethanol:water 25:75-70:30, propylene glycol, or methanol; slightly soluble in acetone, 100% ethanol, chloroform and water; insoluble in 0.1M NaOH. Long Term Storage: +4°C.
Catalog No. 50-200-9520