Learn More
Cliawaived Inc NicCheck 1 Test Strips
SDP
Catalog No. NC1007939
Encompass
(50/Bottle) The only FDA Cleared, CLIAwaived test for Nicotine. NicCheck I Test Strips may be used to detect nicotine and/or its metabolites in urine as an aid in indicating the smoking status of the individual and in planning appropriate treatment. The test will also aid in the identification of a smoker as a low or high nicotine consumer. The NicCheck I test is FDA Cleared and CLIA waived. Individuals, such as physicians, nurses, pharma-cists, medical technologists, and paramedical personnel may perform the test. Each kit includes 50 strips per bottle, 50 tubes, 50 droppers and 1 workstation. Controls sold seperately. Read results in 15 minutes. FDA Cleared and CLIAwaived
Catalog No. NC1007939 Supplier Cliawaived Inc Supplier No. MA50001KIT
(50/Bottle) The only FDA Cleared, CLIAwaived test for Nicotine. NicCheck I Test Strips may be used to detect nicotine and/or its metabolites in urine as an aid in indicating the smoking status of the individual and in planning appropriate treatment. The test will also aid in the identification of a smoker as a low or high nicotine consumer. The NicCheck I test is FDA Cleared and CLIA waived. Individuals, such as physicians, nurses, pharma-cists, medical technologists, and paramedical personnel may perform the test. Each kit includes 50 strips per bottle, 50 tubes, 50 droppers and 1 workstation. Controls sold seperately. Read results in 15 minutes. FDA Cleared and CLIAwaived