Learn More
Sigma Aldrich Fine Chemicals Biosciences Nichipet Eco pipette volum1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7101991EA
Encompass
Providing the accuracy and precision of a micropipette with the compatibility of glass. Glass tips allow pipetting of substances not compatibile with plastic.Glass tip enables totally bias free liquid handling for endocrine disruptors analysis4 sizes cover a wide volume range (20-10000 muL)Nozzle with double O-rings enables easy tip fitting/ejectionReliable sealing nozzle is PTfE and piston is ceramic
Catalog No. 50-290-5575