Learn More
Nichiryo America Inc NICHIPET EX PLUS II 1-10ML
Catalog No. NC1962118
Encompass_Preferred
Autoclavable/UV stable, one touch calibration digital micropipette 1-10ML
Catalog No. NC1962118 Supplier Nichiryo America Inc Supplier No. 00NPL0210ML
Autoclavable/UV stable, one touch calibration digital micropipette 1-10ML