Learn More
Nichiryo America Inc NICHIPET EX PLUS II 10-100ul
Catalog No. NC3373110
Encompass_Preferred
Autoclavable/UV stable, one-touch calibration digital micropipette 10-100ul
Catalog No. NC3373110 Supplier Nichiryo America Inc Supplier No. 00NPL02100
Autoclavable/UV stable, one-touch calibration digital micropipette 10-100ul