Learn More
Nichiryo America Inc NICHIPET EX PLUS II 2-20ul
Catalog No. NC0732259
Encompass_Preferred
Autoclavable/UV stable, one-touch calibration digital micropipette 2-20ul
Catalog No. NC0732259 Supplier Nichiryo America Inc Supplier No. 00NPL0220
Autoclavable/UV stable, one-touch calibration digital micropipette 2-20ul