Learn More
Sigma Aldrich Fine Chemicals Biosciences Nichiryo pipette tips volu
Catalog No. 502901400
Encompass
RNase and DNase freefit most pipettesPremium packaging featuring stand-up and puncture resistant bagsRacked tips are offered in both hinged and lift-off tops
Catalog No. 50-290-1400 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z717401200EA
RNase and DNase freefit most pipettesPremium packaging featuring stand-up and puncture resistant bagsRacked tips are offered in both hinged and lift-off tops