Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Nichrome/Aluminum Inoculating Loops
SCI_ED

Catalog No. S35887 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S35887 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S35887
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fisherbrand Nichrome/Aluminum Inoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. Inoculating loops are used to introduce microorganisms into a culture medium.

  • Fisherbrand™ Nichrome/Aluminum Inoculating Loops are provided with a standard aluminum handle
  • The aluminum handle is available in either an insulated or non-insulated format
  • Inoculating loops are available in a variety of gauges, loop-hole sizes, and wire types to accommodate an array of laboratory requirements.
  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Loops slide along agar surfaces without cutting or tearing material/sample
  • Cools rapidly after sterilization
  • Handle length: 8in.(203mm)
  • Passivated
  • Autoclavable up to 135°C; Can be sterilized by Eto/gamma radiation
  • Latex-free
  • Produced under ISO 9002, GMP, cGMP guidelines

Specifications

Length (English) 3.93 in.
Length (Metric) 10 cm
Material Platinum Iridium and Nichrome
Product Type Inoculating Loop
Gauge 25 G
Includes Tapered end aluminum handle
Length (Metric) Handle 20.3 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.