Learn More
Electron Microscopy Sciences Nichrome Loops, I.D.: 3.5 mm., 1/EA
SDP
Catalog No. 502090953
Encompass_Preferred
These nichrome loops are the perfect tool for the manipulation of samples before and during sample preparation in microscopy. The nichrome wire's composition is 60 percent Ni, 24 percent Fe Alloy, 16 percent Cr. I.D.: 3.5 mm.
Catalog No. 50-209-0953 Supplier Electron Microscopy Sciences Supplier No. 7092205
These nichrome loops are the perfect tool for the manipulation of samples before and during sample preparation in microscopy. The nichrome wire's composition is 60 percent Ni, 24 percent Fe Alloy, 16 percent Cr. I.D.: 3.5 mm.