Learn More
Sigma Aldrich Fine Chemicals Biosciences Nick Translation Kit
Catalog No. 502905588
Encompass
Kit for radioactive or nonradioactive labeling of DNA by the nick translation method. This method utilizes a combination of DNase and DNA Polymerase to nick one strand of the DNA helix then incorporates labeled nucleotides as the polymerase examines or proofreads the nicked site.
Catalog No. 50-290-5588 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10976776001
Kit for radioactive or nonradioactive labeling of DNA by the nick translation method. This method utilizes a combination of DNase and DNA Polymerase to nick one strand of the DNA helix then incorporates labeled nucleotides as the polymerase examines or proofreads the nicked site.